Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Abcf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001190443.1 | NP_001177372.1 | 10 | 1413 | ||
NM_013853.2 | NP_038881.1 | 10 | 1416 | |||
GenBank mRNA | AF213382.1 | - | 3 | 130 | ||
AK087990.1 | - | 10 | 1318 | |||
AK137233.1 | - | 10 | 1348 | |||
AK147937.1 | - | 10 | 1371 | |||
AK151545.1 | - | 10 | 1372 | |||
BC003300.1 | - | 9 | 1308 | 3583837 |