Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Scd3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_024450.2 | NP_077770.1 | 6 | 1589 | ||
GenBank mRNA | AK078675.1 | - | 6 | 1612 | ||
AK078713.1 | - | 6 | 1610 | |||
AK136277.1 | - | 6 | 1606 | |||
BC118033.1 | - | 6 | 1487 | 40106893 |