Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Heatr5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081179.1 | NP_001074648.1 | 10 | 1604 | ||
GenBank mRNA | AK053037.1 | - | 10 | 1588 | ||
AK079542.1 | - | 10 | 1549 | |||
AK146166.1 | - | 10 | 1586 | |||
AK158478.1 | - | 10 | 1589 | |||
AK164172.1 | - | 3 | 478 | |||
AK220390.1 | - | 7 | 1137 | |||
BC086652.1 | - | 9 | 1540 | 6856641 |