Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Olfr100 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_207673.1 | NP_997556.1 | 1 | 113 | ||
GenBank mRNA | BC111097.1 | - | 1 | 112 | ||
BC127233.1 | - | 1 | 113 | 40054913 | ||
BC127234.1 | - | 1 | 113 | 40054914 |
Gene Symbol: Olfr102 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001011721.2 | NP_001011721.2 | 1 | 168 | ||
GenBank mRNA | BC139114.1 | - | 1 | 168 | 8862136 | |
BC139115.1 | - | 1 | 168 | 8862137 |
Gene Symbol: Olfr103 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_146833.1 | NP_667044.1 | 1 | 113 |