Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Otud5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001290536.1 | NP_001277465.1 | 4 | 1070 | ||
NM_001290537.1 | NP_001277466.1 | 5 | 690 | |||
NM_138604.3 | NP_613070.2 | 4 | 1070 | |||
GenBank mRNA | AF229642.1 | - | 4 | 211 | ||
AK010750.1 | - | 5 | 688 | |||
AK149777.1 | - | 5 | 691 | |||
AK155131.1 | - | 4 | 1046 | |||
AK167226.1 | - | 3 | 383 | |||
AK169921.1 | - | 6 | 656 | |||
AK171874.1 | - | 4 | 811 | |||
AK201886.1 | - | 3 | 163 | |||
AK204980.1 | - | 3 | 163 | |||
BC016529.1 | - | 4 | 1070 | 3594583 | ||
BC051111.1 | - | 4 | 809 | 5253679 |