Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019635.2 | NP_062609.2 | 8 | 1110 | ||
GenBank mRNA | AF271361.1 | - | 8 | 856 | ||
AK002914.1 | - | 3 | 328 | |||
AY058922.1 | - | 8 | 996 | |||
BC037440.1 | - | 6 | 866 | 3585312 | ||
BC049123.1 | - | 8 | 984 | 6813956 | ||
U28726.1 | - | 8 | 989 |