Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Stk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019635.2 | NP_062609.2 | 10 | 1492 | ||
GenBank mRNA | AF271361.1 | - | 10 | 1238 | ||
AK002914.1 | - | 5 | 710 | |||
AK004141.1 | - | 1 | 69 | |||
AY058922.1 | - | 10 | 1378 | |||
BC037440.1 | - | 8 | 1248 | 3585312 | ||
BC049123.1 | - | 10 | 1366 | 6813956 | ||
U28726.1 | - | 10 | 1371 |