Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hs1bp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021429.3 | NP_067404.2 | 2, 3 | 228 | ||
GenBank mRNA | AJ132192.1 | - | 2, 3 | 222 | ||
AK040654.1 | - | 2, 3 | 228 | |||
AK084802.1 | - | 2, 3 | 221 | |||
AK154696.1 | - | 2, 3 | 224 | |||
AK170838.1 | - | 2, 3 | 215 | |||
BC095977.1 | - | 2, 3 | 225 |