Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CDMO-CRO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
See other ZNF266 SIRNA Assays ›
Gene Symbol:
ZNF266
Gene Name:
zinc finger protein 266
Gene Aliases:
HZF1
Chromosome Location:
Chr.19: 9412426 - 9435578 on Build GRCh38
UniGene ID:
Hs.656185
Species:
Human
Species Specific ID (Flybase ID):
-
siRNA ID
s21178
Purification
HPLC
Standard
HPLC-IVR
Size
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
1 nmol
5 nmol
InVivo | 5 nmol
20 nmol
20 nmol
40 nmol
40 nmol
250 nmol
250 nmol
InVivo | 250 nmol
InVivo | 250 nmol
1 umol
1 umol
InVivo | 1 umol
InVivo | 1 umol
10 umol
10 umol
Availability
Inventoried
Catalog #
4392420
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
siRNA Details
More Information
Genomic Map
LOADING...
×
Back To Top
siRNA Details
Gene Symbol: ZNF266
Interrogated Sequence
Translated Protein
Targeted Exon(s)
siRNA Location
IMAGE Clone ID
RefSeq
NM_001271314.1
NP_001258243.1
11
2751
NM_006631.3
NP_006622.2
11
2802
XM_005259716.3
XP_005259773.1
8
2468
XM_006722620.2
XP_006722683.1
11
2534
XM_006722621.2
XP_006722684.1
10
2440
XM_006722622.2
XP_006722685.1
9
2279
XM_006722623.3
XP_006722686.1
8
2214
XM_006722627.2
XP_006722690.1
10
3131
XM_006722630.2
XP_006722693.1
9
2445
XM_011527644.1
XP_011525946.1
10
2450
XM_011527647.1
XP_011525949.1
11
2989
XM_011527648.1
XP_011525950.1
11
2985
XM_011527650.1
XP_011525952.1
11
2788
XM_017026154.1
XP_016881643.1
11
2559
XM_017026155.1
XP_016881644.1
11
2556
XM_017026156.1
XP_016881645.1
12
2859
XM_017026157.1
XP_016881646.1
10
3134
XM_017026158.1
XP_016881647.1
12
2830
XM_017026159.1
XP_016881648.1
12
2852
XM_017026160.1
XP_016881649.1
9
3025
XM_017026161.1
XP_016881650.1
9
2730
XM_017026162.1
XP_016881651.1
10
2606
XM_017026163.1
XP_016881652.1
10
2575
XM_017026164.1
XP_016881653.1
8
2380
XM_017026165.1
XP_016881654.1
11
2549
XM_017026166.1
XP_016881655.1
8
2370
XM_017026167.1
XP_016881656.1
8
2204
XM_017026168.1
XP_016881657.1
9
3030
XM_017026169.1
XP_016881658.1
7
2576
XM_017026170.1
XP_016881659.1
8
2190
XM_017026171.1
XP_016881660.1
7
2125
XM_017026172.1
XP_016881661.1
7
2115
XM_017026173.1
XP_016881662.1
10
2470
GenBank mRNA
AB095928.1
-
5
3587
AK027480.1
-
1
1040
AL833471.1
-
4
1707
AL833503.1
-
11
2481
AY376240.1
-
10
2507
BC017407.1
-
1
745
BC071628.1
-
1
1058
BC112919.1
-
9
2691
7502631
BC126255.1
-
4
1777
8991971
BC143940.1
-
4
1777
BC146837.1
-
5
1647
9056764
X78924.1
-
1
465
Back To Top
More Information
Panther Classification:
Molecular Function -
DNA-binding transcription factor
C2H2 zinc finger transcription factor
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
regulation of transcription, DNA-templated
DNA binding
protein binding
metal ion binding
Back To Top
Related Products
Anti-GAPDH, Mouse Monoclonal 6C5
Anti-β-Actin, Mouse Monoclonal AC-15
TaqMan
®
Gene Expression Cells-to-CT
™
Neon
™
Transfection System 100 µL Kit
Lipofectamine
™
RNAiMAX Transfection Reagent
Invivofectamine 2.0
Lipofectamine
™
2000 Transfection Reagent
Neon
™
Transfection System
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-blue-5c9c5c4749-slhn9:80/100.66.78.111:80.
git-commit: d22414ba47b22cc02ffc24003f0e179da61511a6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.21.0-Offline