Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037637.1 | NP_001032726.1 | 4 | 719 | ||
NM_001207.4 | NP_001198.2 | 4 | 478 | |||
GenBank mRNA | AB062126.1 | - | 4 | 451 | ||
AK225964.1 | - | 4 | 461 | |||
AK291125.1 | - | 4 | 455 | |||
BC008062.2 | - | 4 | 446 | 2966788 | ||
BT007120.1 | - | 3 | 344 | |||
BU661989.1 | - | 4 | 495 | |||
BX359147.2 | - | 4 | 677 | |||
X53280.1 | - | 4 | 610 | |||
X74070.1 | - | 4 | 446 |