Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF274 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278734.1 | NP_001265663.1 | 8 | 2009 | ||
NM_016324.3 | NP_057408.2 | 5 | 1693 | |||
NM_016325.3 | NP_057409.1 | 7 | 1898 | |||
NM_133502.2 | NP_598009.1 | 8 | 1994 | |||
XM_011526327.1 | XP_011524629.1 | 7 | 1432 | |||
XM_011526328.2 | XP_011524630.1 | 3 | 1113 | |||
XM_017026174.1 | XP_016881663.1 | 6 | 1699 | |||
XM_017026175.1 | XP_016881664.1 | 6 | 1854 | |||
GenBank mRNA | AB029149.1 | - | 5 | 1653 | ||
AB029150.1 | - | 7 | 1800 | |||
AF275680.1 | - | 8 | 1673 | |||
AF275681.1 | - | 2 | 1107 | |||
AK095162.1 | - | 2 | 2338 | |||
AK222855.1 | - | 5 | 1621 | |||
AK222878.1 | - | 5 | 1621 | |||
BC009763.2 | - | 7 | 1521 | 2966764 | ||
BT007304.1 | - | 7 | 1495 | |||
BX641169.1 | - | 2 | 4108 |