Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ralb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022327.5 | NP_071722.1 | 3 | 424 | ||
GenBank mRNA | AF174296.1 | - | 392 | |||
AK017698.1 | - | 3 | 270 | |||
AK033193.1 | - | 3 | 279 | |||
AK159993.1 | - | 3 | 369 | |||
AK205791.1 | - | 3 | 238 | |||
BC006907.1 | - | 3 | 268 | 3599947 |