Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF274 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001278734.1 | NP_001265663.1 | 8 | 2002 | ||
NM_016324.3 | NP_057408.2 | 5 | 1686 | |||
NM_016325.3 | NP_057409.1 | 7 | 1891 | |||
NM_133502.2 | NP_598009.1 | 8 | 1987 | |||
XM_011526327.1 | XP_011524629.1 | 7 | 1425 | |||
XM_011526328.2 | XP_011524630.1 | 3 | 1106 | |||
XM_017026174.1 | XP_016881663.1 | 6 | 1692 | |||
XM_017026175.1 | XP_016881664.1 | 6 | 1847 | |||
GenBank mRNA | AB029149.1 | - | 5 | 1646 | ||
AB029150.1 | - | 7 | 1793 | |||
AF275680.1 | - | 8 | 1666 | |||
AF275681.1 | - | 2 | 1100 | |||
AK095162.1 | - | 2 | 2331 | |||
AK222855.1 | - | 5 | 1614 | |||
AK222878.1 | - | 5 | 1614 | |||
BC009763.2 | - | 7 | 1514 | 2966764 | ||
BT007304.1 | - | 7 | 1488 | |||
BX641169.1 | - | 2 | 4101 |