Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gpr89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026229.4 | NP_080505.1 | 7 | 674 | ||
GenBank mRNA | AK002937.1 | - | 7 | 694 | ||
AK009734.1 | - | 6 | 553 | |||
AK016787.1 | - | 7 | 674 | |||
AK034931.1 | - | 242 | ||||
BC010729.1 | - | 7 | 667 | 3708143 |
Set Membership: |
G-protein coupled receptor |