Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ssna1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_023464.2 | NP_075953.1 | 3 | 481 | ||
GenBank mRNA | AB041656.1 | - | 3 | 475 | ||
AK003356.2 | - | 3 | 482 | |||
AK004482.1 | - | 1 | 360 | |||
AK088418.1 | - | 3 | 413 | |||
AK158412.1 | - | 598 | ||||
AK208309.1 | - | 2 | 173 | |||
BC023033.1 | - | 3 | 444 | |||
BC037081.1 | - | 3 | 458 | 5098653 | ||
BC048360.1 | - | 3 | 459 | 5067090 | ||
BY703613.1 | - | 3 | 482 |