Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ace2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130513.1 | NP_001123985.1 | 9 | 1314 | ||
NM_027286.4 | NP_081562.2 | 8 | 1166 | |||
GenBank mRNA | AB053181.1 | - | 8 | 1170 | ||
AB053182.1 | - | 8 | 1170 | |||
AK135357.1 | - | 3 | 348 | |||
AK141604.1 | - | 8 | 1015 | |||
BC026801.1 | - | 8 | 1159 | 4236529 | ||
EF408739.1 | - | 9 | 1275 | |||
EF408740.1 | - | 9 | 1275 | |||
GQ262785.1 | - | 8 | 984 |