Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: IQGAP2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006633.3 | NP_006624.2 | 1, 2 | 343 | ||
XM_017008960.1 | XP_016864449.1 | 1, 2 | 292 | |||
GenBank mRNA | AK291066.1 | - | 1, 2 | 343 | ||
U51903.1 | - | 1, 2 | 274 |