Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1galt1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_052993.3 | NP_443719.3 | 3 | 926 | ||
GenBank mRNA | AF157962.1 | - | 3 | 886 | ||
BC025899.1 | - | 2 | 549 | |||
BC064767.1 | - | 3 | 817 | 30077842 | ||
BG077752.2 | - | 1 | 338 |