Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF273 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021148.2 | NP_066971.2 | 4 | 1701 | ||
NR_003099.1 | 5 | 1790 | ||||
XM_011515746.2 | XP_011514048.1 | 3 | 1636 | |||
XM_017011688.1 | XP_016867177.1 | 4 | 1727 | |||
XM_017011689.1 | XP_016867178.1 | 9 | 7715 | |||
XM_017011690.1 | XP_016867179.1 | 8 | 7626 | |||
XM_017011691.1 | XP_016867180.1 | 9 | 4917 | |||
XM_017011692.1 | XP_016867181.1 | 7 | 7101 | |||
XM_017011693.1 | XP_016867182.1 | 8 | 2807 | |||
XM_017011694.1 | XP_016867183.1 | 8 | 2165 | |||
XM_017011695.1 | XP_016867184.1 | 5 | 1776 | |||
XM_017011696.1 | XP_016867185.1 | 6 | 2002 | |||
XM_017011697.1 | XP_016867186.1 | 7 | 7537 | |||
XM_017011698.1 | XP_016867187.1 | 6 | 2654 | |||
GenBank mRNA | AK090648.1 | - | 8 | 2808 | ||
AL832810.1 | - | 1 | 1364 | |||
BC063818.1 | - | 5 | 1756 | 6503687 | ||
X78932.1 | - | 1 | 661 |