Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ZNF268 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165881.2 | NP_001159353.1 | 6 | 985 | ||
NM_001165882.2 | NP_001159354.1 | 6 | 992 | |||
NM_001165883.1 | NP_001159355.1 | 4 | 886 | |||
NM_001165884.2 | NP_001159356.2 | 3 | 685 | |||
NM_001165885.1 | NP_001159357.1 | 5 | 982 | |||
NM_001165886.1 | NP_001159358.1 | 4 | 781 | |||
NM_001165887.1 | NP_001159359.1 | 6 | 1014 | |||
NM_003415.2 | NP_003406.1 | 6 | 1109 | |||
NM_152943.2 | NP_694422.2 | 7 | 1215 | |||
GenBank mRNA | AF317549.1 | - | 6 | 1109 | ||
AF385187.1 | - | 7 | 1065 | |||
AF432217.1 | - | 3 | 585 | |||
AK027712.1 | - | 7 | 1129 | |||
AY583691.1 | - | 8 | 961 | |||
BC110542.2 | - | 6 | 985 | 40036577 |