Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RPP40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001286132.1 | NP_001273061.1 | 4 | 461 | ||
NM_006638.3 | NP_006629.2 | 5 | 530 | |||
XM_011514253.2 | XP_011512555.1 | 5 | 525 | |||
GenBank mRNA | BC017871.1 | - | 4 | 435 | 4278825 | |
BG430780.1 | - | 5 | 505 | |||
BM015573.1 | - | 5 | 562 | |||
BQ220155.1 | - | 5 | 478 |