Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001731.2 | NP_001722.1 | 2 | 780 | ||
GenBank mRNA | AK307057.1 | - | 3 | 533 | ||
AK313309.1 | - | 2 | 731 | |||
BC016759.2 | - | 2 | 722 | 2987983 | ||
BC064953.1 | - | 2 | 723 | 6050290 | ||
BT019839.1 | - | 2 | 418 | |||
BT019840.1 | - | 2 | 418 | |||
BT019841.1 | - | 2 | 418 | |||
CR456876.1 | - | 2 | 418 | |||
X61123.1 | - | 2 | 726 |