Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SDCCAG3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039707.1 | NP_001034796.1 | 10 | 1467 | ||
NM_001039708.1 | NP_001034797.1 | 8 | 1248 | |||
NM_006643.3 | NP_006634.3 | 9 | 1398 | |||
XM_005266050.3 | XP_005266107.1 | 10 | 1394 | |||
XM_005266051.3 | XP_005266108.1 | 9 | 1312 | |||
XM_011518157.2 | XP_011516459.1 | 9 | 1251 | |||
XM_017014218.1 | XP_016869707.1 | 9 | 1568 | |||
XM_017014219.1 | XP_016869708.1 | 9 | 1541 | |||
GenBank mRNA | AF039688.1 | - | 6 | 535 | ||
AK001296.1 | - | 8 | 1027 | |||
AY129014.1 | - | 3 | 1341 | |||
AY349357.1 | - | 9 | 1088 | |||
BC014515.1 | - | 9 | 1265 | 3547130 |