Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STARD10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006645.2 | NP_006636.2 | 2 | 1076 | ||
GenBank mRNA | AF151810.1 | - | 2 | 612 | ||
AK095668.1 | - | 2 | 992 | |||
AK310301.1 | - | 3 | 601 | |||
AK315255.1 | - | 3 | 870 | |||
BC007919.2 | - | 3 | 595 | 4301295 | ||
BC014033.2 | - | 3 | 595 | 4330790 |