Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000061.2 | NP_000052.1 | 5, 6 | 577 | ||
NM_001287344.1 | NP_001274273.1 | 5, 6 | 733 | |||
NM_001287345.1 | NP_001274274.1 | 6, 7 | 742 | |||
GenBank mRNA | AK057105.1 | - | 6, 7 | 696 | ||
AK225577.1 | - | 5, 6 | 539 | |||
AK309851.1 | - | 5, 6 | 719 | |||
AK314382.1 | - | 5, 6 | 544 | |||
BC109079.1 | - | 5, 6 | 509 | 40034287 | ||
BC109080.1 | - | 5, 6 | 509 | 40034289 | ||
KF241986.1 | - | 4, 5 | 384 | |||
X58957.1 | - | 5, 6 | 516 |