Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BTK | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_000061.2 | NP_000052.1 | 3 | 351 | ||
NM_001287344.1 | NP_001274273.1 | 3 | 507 | |||
NM_001287345.1 | NP_001274274.1 | 4 | 516 | |||
GenBank mRNA | AF153755.1 | - | 2 | 158 | ||
AK057105.1 | - | 4 | 470 | |||
AK225577.1 | - | 3 | 313 | |||
AK309851.1 | - | 3 | 493 | |||
AK314382.1 | - | 3 | 318 | |||
BC109079.1 | - | 3 | 283 | 40034287 | ||
BC109080.1 | - | 3 | 283 | 40034289 | ||
DA619542.1 | - | 3 | 351 | |||
KF241986.1 | - | 2 | 158 | |||
X58957.1 | - | 3 | 290 |