Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: FAXDC2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032385.4 | NP_115761.2 | 9 | 1944 | ||
XM_005268358.2 | XP_005268415.1 | 6 | 1589 | |||
XM_006714753.2 | XP_006714816.1 | 10 | 2319 | |||
XM_017008965.1 | XP_016864454.1 | 10 | 2344 | |||
XM_017008966.1 | XP_016864455.1 | 7 | 1619 | |||
GenBank mRNA | AF159165.1 | - | 1 | 1950 | ||
AK023820.1 | - | 8 | 2735 | |||
AK074245.1 | - | 3 | 1089 | |||
AL359655.1 | - | 2 | 387 | |||
BC004506.2 | - | 7 | 1641 | |||
BC007216.1 | - | 9 | 1912 | 2906244 |