Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ALDH1L1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001270364.1 | NP_001257293.1 | 4 | 625 | ||
NM_012190.3 | NP_036322.2 | 4 | 727 | |||
NR_072979.1 | 4 | 727 | ||||
XM_006713481.2 | XP_006713544.1 | 4 | 466 | |||
XM_011512355.1 | XP_011510657.1 | 4 | 459 | |||
XM_017005613.1 | XP_016861102.1 | 4 | 850 | |||
XM_017005614.1 | XP_016861103.1 | 4 | 466 | |||
GenBank mRNA | AB209299.1 | - | 4 | 534 | ||
AF052732.1 | - | 4 | 487 | |||
AK222535.1 | - | 4 | 509 | |||
AK222694.1 | - | 4 | 509 | |||
AK294530.1 | - | 3 | 276 | |||
AK307699.1 | - | 5 | 638 | |||
AK308112.1 | - | 4 | 466 | |||
AL133015.1 | - | 2 | 100 | |||
BC027241.1 | - | 4 | 500 | 4484301 | ||
BI829048.1 | - | 4 | 568 | |||
CR749807.1 | - | 4 | 625 | |||
KM111188.1 | - | 4 | 536 | |||
KU178540.1 | - | 3 | 377 | |||
KU178541.1 | - | 4 | 561 | |||
KU178542.1 | - | 3 | 377 |