Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PDE10A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001130690.2 | NP_001124162.1 | 18 | 2039 | ||
NM_006661.3 | NP_006652.1 | 19 | 2167 | |||
XM_006715321.3 | XP_006715384.1 | 18 | 3364 | |||
XM_011535387.2 | XP_011533689.2 | 20 | 4159 | |||
XM_011535388.2 | XP_011533690.1 | 19 | 3492 | |||
XM_011535393.2 | XP_011533695.1 | 18 | 1967 | |||
XM_017010194.1 | XP_016865683.1 | 20 | 4159 | |||
XM_017010195.1 | XP_016865684.1 | 18 | 1912 | |||
XM_017010196.1 | XP_016865685.1 | 20 | 3610 | |||
GenBank mRNA | AB020593.1 | - | 19 | 2172 | ||
AB026816.1 | - | 18 | 1972 | |||
AF127479.1 | - | 18 | 1988 | |||
AF127480.1 | - | 18 | 2008 | |||
AK021956.1 | - | 2 | 264 | |||
AK297555.1 | - | 9 | 1118 | |||
BC104858.1 | - | 18 | 1959 | 8143861 | ||
BC104860.1 | - | 18 | 1959 | 8143863 | ||
BC143297.1 | - | 19 | 2078 | |||
BC143301.1 | - | 19 | 2078 | |||
CR536567.1 | - | 18 | 1922 | |||
KU232567.1 | - | 18 | 2410 |