Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SRCAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006662.2 | NP_006653.2 | 11 | 1800 | ||
GenBank mRNA | AB621816.1 | - | 11 | 1867 | ||
AF143946.1 | - | 7 | 1034 | |||
AK023808.1 | - | 4 | 514 | |||
AK128030.1 | - | 11 | 1800 | |||
AU140541.2 | - | 4 | 514 | |||
BC025284.1 | - | 8 | 1277 | |||
BC063578.1 | - | 9 | 1594 |