Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PPP1R13L | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001142502.1 | NP_001135974.1 | 13 | 3000 | ||
NM_006663.3 | NP_006654.2 | 13 | 2998 | |||
XM_017026177.1 | XP_016881666.1 | 14 | 3056 | |||
XM_017026178.1 | XP_016881667.1 | 14 | 3116 | |||
GenBank mRNA | AF078036.1 | - | 7 | 1979 | ||
AF078037.1 | - | 6 | 2433 | |||
AY869712.1 | - | 13 | 2979 | |||
BC001475.2 | - | 7 | 1585 | |||
BC020589.1 | - | 6 | 1239 | |||
BC032298.1 | - | 13 | 2987 | 4994121 | ||
BC064913.1 | - | 13 | 2963 | 6068949 |