Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ADAM28 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001304351.1 | NP_001291280.1 | 11 | 1168 | ||
NM_014265.5 | NP_055080.2 | 11 | 1168 | |||
NM_021777.4 | NP_068547.2 | 11 | 1168 | |||
NR_130709.1 | 10 | 975 | ||||
NR_130710.1 | 11 | 1230 | ||||
XM_005273380.3 | XP_005273437.1 | 11 | 1168 | |||
XM_005273382.1 | XP_005273439.1 | 11 | 1168 | |||
XM_006716273.3 | XP_006716336.1 | 11 | 1168 | |||
XM_006716274.1 | XP_006716337.1 | 11 | 1168 | |||
XM_011544367.2 | XP_011542669.1 | 11 | 1168 | |||
XM_011544368.2 | XP_011542670.1 | 11 | 1168 | |||
XM_011544369.2 | XP_011542671.1 | 11 | 1168 | |||
XM_011544370.2 | XP_011542672.1 | 10 | 946 | |||
XM_011544371.2 | XP_011542673.1 | 4 | 390 | |||
XM_017012974.1 | XP_016868463.1 | 11 | 1168 | |||
XM_017012975.1 | XP_016868464.1 | 11 | 1168 | |||
XM_017012976.1 | XP_016868465.1 | 12 | 1267 | |||
GenBank mRNA | AF137334.1 | - | 11 | 1097 | ||
AF137335.1 | - | 11 | 1097 | |||
AJ242015.1 | - | 11 | 1105 | |||
AK225921.1 | - | 1915 | ||||
AK293382.1 | - | 10 | 947 | |||
AK303437.1 | - | 11 | 1201 | |||
BC136478.1 | - | 11 | 1084 | 9020467 |