Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: SLC22A7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006672.3 | NP_006663.2 | 2 | 518 | ||
NM_153320.2 | NP_696961.2 | 3 | 524 | |||
XM_006714970.3 | XP_006715033.1 | 3 | 524 | |||
XM_006714971.3 | XP_006715034.1 | 2 | 518 | |||
XM_011514256.2 | XP_011512558.1 | 3 | 701 | |||
XM_011514257.2 | XP_011512559.1 | 3 | 701 | |||
XM_011514259.1 | XP_011512561.1 | 3 | 701 | |||
XM_011514261.2 | XP_011512563.1 | 4 | 466 | |||
XM_011514262.2 | XP_011512564.1 | 3 | 701 | |||
XM_011514263.2 | XP_011512565.1 | 3 | 277 | |||
XM_017010198.1 | XP_016865687.1 | 3 | 701 | |||
XM_017010199.1 | XP_016865688.1 | 3 | 701 | |||
XM_017010200.1 | XP_016865689.1 | 4 | 506 | |||
XM_017010201.1 | XP_016865690.1 | 4 | 476 | |||
GenBank mRNA | AF097518.1 | - | 3 | 520 | ||
AF210455.1 | - | 2 | 419 | |||
AK290796.1 | - | 2 | 437 | |||
AK290806.1 | - | 2 | 496 | |||
AY050498.1 | - | 3 | 425 | |||
BC017963.1 | - | 2 | 731 | |||
BC033805.1 | - | 3 | 445 | 5187472 | ||
DA640137.1 | - | 3 | 524 |