Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TSPAN9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168320.1 | NP_001161792.1 | 8 | 1382 | ||
NM_006675.4 | NP_006666.1 | 9 | 1449 | |||
XM_011520912.2 | XP_011519214.1 | 10 | 1566 | |||
XM_011520913.2 | XP_011519215.1 | 7 | 1308 | |||
GenBank mRNA | AK027366.1 | - | 9 | 1425 | ||
AK223179.1 | - | 9 | 1421 | |||
AY523992.1 | - | 1 | 212 |