Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BYSL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004053.3 | NP_004044.3 | 4, 5 | 1045 | ||
GenBank mRNA | AK095253.1 | - | 1465 | |||
AK314147.1 | - | 4, 5 | 800 | |||
BC007340.2 | - | 4, 5 | 781 | 2989807 | ||
BC050645.1 | - | 4, 5 | 1045 | 6011459 | ||
BC062627.1 | - | 4, 5 | 1016 | 5226210 | ||
L36720.2 | - | 3, 4 | 325 |