Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: CD300C | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006678.4 | NP_006669.1 | 2 | 583 | ||
XM_017024033.1 | XP_016879522.1 | 2 | 578 | |||
XM_017024034.1 | XP_016879523.1 | 2 | 578 | |||
GenBank mRNA | BC022279.1 | - | 2 | 555 | 4692025 | |
X66171.1 | - | 2 | 433 |