Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BYSL | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_004053.3 | NP_004044.3 | 3, 4 | 910 | ||
GenBank mRNA | AK095253.1 | - | 1330 | |||
AK314147.1 | - | 3, 4 | 665 | |||
BC007340.2 | - | 3, 4 | 646 | 2989807 | ||
BC050645.1 | - | 3, 4 | 910 | 6011459 | ||
BC062627.1 | - | 3, 4 | 881 | 5226210 | ||
L36720.2 | - | 2, 3 | 190 |