Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1QBP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001212.3 | NP_001203.1 | 5, 6 | 785 | ||
GenBank mRNA | AF275902.1 | - | 4, 5 | 452 | ||
AK291516.1 | - | 5, 6 | 745 | |||
BC000435.2 | - | 5, 6 | 739 | 2820039 | ||
BC013731.1 | - | 5, 6 | 728 | 3862881 | ||
BT019898.1 | - | 5, 6 | 707 | |||
BT019899.1 | - | 5, 6 | 707 | |||
L04636.1 | - | 5, 6 | 785 | |||
X75913.1 | - | 5, 6 | 785 |