Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: MALT1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_006785.3 | NP_006776.1 | 16 | 2261 | ||
NM_173844.2 | NP_776216.1 | 15 | 2228 | |||
GenBank mRNA | AB026118.1 | - | 15 | 2035 | ||
AF130356.2 | - | 16 | 2167 | |||
AF316597.1 | - | 16 | 2261 | |||
AK024859.1 | - | 4 | 453 | |||
AK092004.1 | - | 5 | 1585 | |||
AK291386.1 | - | 16 | 2227 | |||
AL137399.1 | - | 2 | 213 | |||
BC030143.2 | - | 15 | 2082 | 4765195 |