Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: RUNDC3A | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001144825.1 | NP_001138297.1 | 4, 5 | 734 | ||
NM_001144826.1 | NP_001138298.1 | 4, 5 | 719 | |||
NM_006695.4 | NP_006686.1 | 4, 5 | 734 | |||
XM_005256964.3 | XP_005257021.1 | 4, 5 | 667 | |||
XM_017024035.1 | XP_016879524.1 | 4, 5 | 734 | |||
XM_017024036.1 | XP_016879525.1 | 4, 5 | 655 | |||
XM_017024037.1 | XP_016879526.1 | 4, 5 | 667 | |||
XM_017024038.1 | XP_016879527.1 | 4, 5 | 734 | |||
XM_017024039.1 | XP_016879528.1 | 4, 5 | 734 | |||
XM_017024040.1 | XP_016879529.1 | 4, 5 | 655 | |||
GenBank mRNA | AB209802.1 | - | 4, 5 | 718 | ||
AF055026.1 | - | 4, 5 | 498 | |||
AK226045.1 | - | 4, 5 | 695 | |||
AK313669.1 | - | 4, 5 | 655 | |||
AL133657.1 | - | 4, 5 | 880 | |||
BC006194.2 | - | 4, 5 | 697 | 2823366 | ||
BX537443.1 | - | 4, 5 | 616 | |||
CR456748.1 | - | 4, 5 | 460 | |||
U93871.1 | - | 4, 5 | 445 |