Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: DHRS4L2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001193636.1 | NP_001180565.1 | 3, 4 | 743 | ||
NM_198083.3 | NP_932349.2 | 3, 4 | 542 | |||
GenBank mRNA | BC000663.2 | - | 3, 4 | 450 | 3349226 | |
DQ088987.1 | - | 1, 2 | 41 | |||
DQ088988.1 | - | 1, 2 | 41 | |||
GU564235.1 | - | 3, 4 | 510 |
Gene Symbol: DHRS4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021004.3 | NP_066284.2 | 3, 4 | 466 | ||
XM_006720005.3 | XP_006720068.1 | 3, 4 | 466 | |||
GenBank mRNA | AB045131.1 | - | 3, 4 | 491 | ||
AF044127.1 | - | 3, 4 | 437 | |||
AF064256.1 | - | 3, 4 | 440 | |||
AK001870.1 | - | 3, 4 | 432 | |||
AK314448.1 | - | 3, 4 | 431 | |||
AY358638.1 | - | 3, 4 | 447 | |||
BC003019.1 | - | 3, 4 | 414 | 2822884 | ||
BP366505.1 | - | 3, 4 | 470 | |||
DQ344810.1 | - | 3, 4 | 445 |