Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: BLCAP | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167820.1 | NP_001161292.1 | 3 | 614 | ||
NM_001167821.1 | NP_001161293.1 | 2 | 444 | |||
NM_001167822.2 | NP_001161294.1 | 2 | 501 | |||
NM_001167823.1 | NP_001161295.1 | 2 | 517 | |||
NM_001317074.1 | NP_001304003.1 | 2 | 494 | |||
NM_001317075.1 | NP_001304004.1 | 2 | 410 | |||
NM_006698.3 | NP_006689.1 | 2 | 481 | |||
XM_017027599.1 | XP_016883088.1 | 2 | 1394 | |||
GenBank mRNA | AF053470.1 | - | 2 | 420 | ||
AK074498.1 | - | 4 | 511 | |||
AK124822.1 | - | 3 | 339 | |||
AL049288.1 | - | 2 | 427 | |||
BC047692.1 | - | 2 | 412 | 6059195 | ||
BC071704.1 | - | 2 | 406 | 5190373 | ||
BP293276.1 | - | 2 | 517 | |||
CB155706.1 | - | 2 | 451 | |||
DA198124.1 | - | 2 | 444 | |||
DA262965.1 | - | 2 | 481 | |||
DA283758.1 | - | 2 | 410 | |||
DA632207.1 | - | 3 | 563 | |||
DA770824.1 | - | 2 | 481 | |||
DA943717.1 | - | 2 | 450 |