Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1QA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015991.2 | NP_057075.1 | 3 | 551 | ||
GenBank mRNA | AF135157.1 | - | 2 | 478 | ||
AF260332.1 | - | 2 | 483 | |||
AK311980.1 | - | 3 | 494 | |||
BC030153.2 | - | 3 | 521 | 4850418 | ||
BC071986.1 | - | 2 | 487 | 6300836 |