Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: PAPOLA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001293627.1 | NP_001280556.1 | 17 | 1790 | ||
NM_001293628.1 | NP_001280557.1 | 17 | 1655 | |||
NM_001293632.1 | NP_001280561.1 | 15 | 1626 | |||
NM_032632.4 | NP_116021.2 | 17 | 1790 | |||
XM_005267281.1 | XP_005267338.1 | 17 | 1786 | |||
XM_005267282.2 | XP_005267339.1 | 17 | 1786 | |||
XM_017020935.1 | XP_016876424.1 | 17 | 1786 | |||
XM_017020936.1 | XP_016876425.1 | 17 | 1786 | |||
GenBank mRNA | AK024849.1 | - | 3 | 907 | ||
AK295020.1 | - | 17 | 1621 | |||
AK302406.1 | - | 18 | 1651 | |||
AK302984.1 | - | 5 | 442 | |||
AK303402.1 | - | 4 | 342 | |||
AK307659.1 | - | 15 | 1585 | |||
AK307680.1 | - | 7 | 638 | |||
AK316168.1 | - | 2 | 149 | |||
BC036014.1 | - | 17 | 1785 | 5271668 | ||
BX248301.1 | - | 17 | 1655 | |||
X76770.1 | - | 16 | 1549 |