Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: TCERG1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040006.1 | NP_001035095.1 | 13 | 2035 | ||
NM_006706.3 | NP_006697.2 | 14 | 2098 | |||
XM_005268365.2 | XP_005268422.1 | 15 | 2335 | |||
XM_017008977.1 | XP_016864466.1 | 14 | 2272 | |||
XM_017008978.1 | XP_016864467.1 | 15 | 2335 | |||
XM_017008979.1 | XP_016864468.1 | 14 | 2097 | |||
XM_017008980.1 | XP_016864469.1 | 15 | 2335 | |||
XM_017008981.1 | XP_016864470.1 | 12 | 1690 | |||
XM_017008982.1 | XP_016864471.1 | 14 | 2066 | |||
XM_017008983.1 | XP_016864472.1 | 13 | 2015 | |||
GenBank mRNA | AB209910.1 | - | 13 | 2009 | ||
AF017789.1 | - | 14 | 2058 | |||
AK126455.1 | - | 12 | 1938 | |||
BC014151.2 | - | 13 | 2029 | |||
BC111727.1 | - | 13 | 2013 | 40028766 |