Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: EHMT2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001289413.1 | NP_001276342.1 | 7 | 1060 | ||
NM_001318833.1 | NP_001305762.1 | 8 | 1144 | |||
NM_006709.4 | NP_006700.3 | 8 | 915 | |||
NM_025256.6 | NP_079532.5 | 8 | 915 | |||
XM_005248824.3 | XP_005248881.1 | 7 | 1056 | |||
XM_006714974.1 | XP_006715037.1 | 7 | 1060 | |||
XM_006714975.1 | XP_006715038.1 | 7 | 1060 | |||
XM_006714976.1 | XP_006715039.1 | 8 | 898 | |||
XM_017010202.1 | XP_016865691.1 | 8 | 898 | |||
GenBank mRNA | AB209433.2 | - | 7 | 977 | ||
AJ315532.1 | - | 8 | 885 | |||
AK302904.1 | - | 7 | 1060 | |||
BC002686.1 | - | 5 | 385 | |||
BC009351.1 | - | 8 | 882 | |||
BC018718.1 | - | 8 | 873 | 4123580 | ||
BC020970.1 | - | 7 | 850 | |||
DA099526.1 | - | 5 | 525 | |||
X69838.1 | - | 4 | 305 |