Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C1QC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001114101.1 | NP_001107573.1 | 3 | 489 | ||
NM_172369.3 | NP_758957.2 | 3 | 466 | |||
GenBank mRNA | AF087892.1 | - | 3 | 464 | ||
AK057792.1 | - | 3 | 481 | |||
AK130613.1 | - | 3 | 464 | |||
AK226152.1 | - | 1 | 3761 | |||
BC009016.1 | - | 3 | 431 | 4212772 | ||
CB995661.1 | - | 3 | 516 | |||
DA849505.1 | - | 3 | 458 |