Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: POP1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001145860.1 | NP_001139332.1 | 4 | 477 | ||
NM_001145861.1 | NP_001139333.1 | 4 | 417 | |||
NM_015029.2 | NP_055844.2 | 4 | 405 | |||
XM_011516801.2 | XP_011515103.1 | 4 | 477 | |||
GenBank mRNA | AK291434.1 | - | 4 | 474 | ||
BC011529.1 | - | 4 | 405 | 3853321 | ||
BP249298.1 | - | 4 | 403 | |||
BP280616.1 | - | 4 | 477 | |||
D31765.1 | - | 1 | 16 |