Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: C11orf58 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_014267.5 | NP_055082.1 | 2 | 314 | ||
XM_017017143.1 | XP_016872632.1 | 2 | 2573 | |||
GenBank mRNA | AK315382.1 | - | 2 | 268 | ||
BC007103.1 | - | 2 | 268 | 4281282 | ||
BC016352.1 | - | 2 | 272 | 4082845 | ||
BC020937.1 | - | 2 | 268 | 4281930 | ||
BC103747.1 | - | 2 | 295 | 6151644 | ||
BP210918.1 | - | 3 | 352 | |||
BP226882.1 | - | 2 | 514 | |||
BU661194.1 | - | 3 | 363 | |||
CR456882.1 | - | 2 | 136 | |||
U51678.1 | - | 2 | 171 |