Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: STARD3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001165937.1 | NP_001159409.1 | 9 | 976 | ||
NM_001165938.1 | NP_001159410.1 | 8 | 922 | |||
NM_006804.3 | NP_006795.3 | 9 | 976 | |||
XM_006721646.2 | XP_006721709.1 | 8 | 846 | |||
XM_011524210.1 | XP_011522512.1 | 9 | 945 | |||
XM_011524211.2 | XP_011522513.1 | 9 | 948 | |||
XM_011524213.1 | XP_011522515.1 | 8 | 846 | |||
XM_017024041.1 | XP_016879530.1 | 9 | 948 | |||
GenBank mRNA | AB209772.1 | - | 3 | 502 | ||
AK123364.1 | - | 2 | 189 | |||
AK225069.1 | - | 9 | 909 | |||
AK300176.1 | - | 8 | 858 | |||
AK300842.1 | - | 9 | 866 | |||
AK303114.1 | - | 8 | 847 | |||
AL831952.1 | - | 3 | 1037 | |||
BC008356.1 | - | 9 | 927 | 3547271 | ||
BC008747.2 | - | 9 | 896 | 3346043 | ||
BC025679.1 | - | 9 | 897 | 5203654 | ||
BT006964.1 | - | 8 | 773 | |||
D38255.1 | - | 9 | 894 | |||
X80198.1 | - | 9 | 882 |